Learn More
Micronova™ Circular Stainless-Steel Bucket
SDP
Catalog No. p-3350263 Shop All Micronova Manufacturing Products
Click to view available options
Product Type:
Bucket
Mop Wringer
Corrosion-resistant.
  • Bucket features four 2 in. dia. (5cm) casters and handle for easy mobility and opening dia. of 13.75 in. (34.9cm)
  • Optional downward press, stainless-steel wringer quickly and easily removes excess liquid from mop
Specifications
Product Type Bucket
Capacity (Metric) 30.3 L
Material Stainless Steel
Autoclavable Autoclavable
Capacity (English) 32 qt.
For Use With (Application) Cleanroom and cleanroom support for floors and walls
Includes Four 2 in. dia. casters and handle
Quantity 1 Ea.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.