Learn More
Micronova™ MegaClean™ Heavy-Duty Cleaner
BIOPHARMASDP
Catalog No. 19035380e
Click to view available options
Volume (English):
1 gal.
5 gal.
A non-flammable, heavy duty cleaner and degreaser that removes grease, adhesives and photo-resist stains from most surfaces. Available in 1 or 5 gallon bottles and a 1 quart spray bottle.
The Micronova MegaClean™ Heavy-Duty Cleaner offers an effective alternative to toxic, flammable, solvent-based cleaners, such as acetone and isopropyl alcohol. Filtered to 0.1 microns, this cleaner is ideal for semiconductor, electronics, aerospace and clean-build operations.
  • Non-flammable, heavy duty cleanroom degreaser
  • Removes grease, adhesives and photo-resist stains from most surfaces
  • Ideal for semiconductor, electronics, aerospace and clean-build operations
  • Filtered to 0.1 microns
  • Available in 1 or 5 gallon refill bottles and a 1 quart spray bottle
  • Biodegradable
  • Water soluble
  • Effective at low concentrations
Specifications
Sterility Non-sterile
Description Heavy Duty Detergent, Degreaser
For Use With (Application) Heavy Soils, Adhesive Removal in and Around Cleanroom
For Use With (Equipment) Mops and Buckets or Wipes and Sponge
Volume (English) 1 gal.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.