Learn More
Micronova™ NovaMop™ Roller Mops
SDP
Catalog No. p-102491
Click to view available options
Material:
NovaLite
Polyester, Urethane Foam
Polyvinyl Acetate
Use to clean floors, walls, and ceilings. Lightweight roller mops with foam interiors and choice of three fabric coverings. Mop heads free roll in one direction and lock on the reverse roll to effectively dry the surfaces being cleaned. The same roll/lock formula allows several clean passes with one mop head.
NovaMop PVA. Ideal for semiconductor and microelectronics facilities. Low linting, fast drying mop leaves cleaned surfaces dry and residue free. Launderable. For Class M1.5 to M6.5 (Class 1 to 100,000) cleanrooms.NovaMop Lite. Offers similar absorbency to NovaMop PVA but holds up to stronger disinfectants and autoclaves. For Class M3.5 to M6.5 (Class 100 to 100,000) cleanrooms.NovaMop Polyester. Designed for sterile applications. Can be autoclaved many times and used with strong disinfectants. Two-ply cover allows the mop to be saturated for optimum wetting and sterilizing purposes. Can be laundered and autoclaved. For Class M2.5 to M6.5 (Class 10 to 100,000) cleanrooms.
Order Info
Quick-disconnect NovaMop adapter sold separately. Extendible and standard stainless-steel handles also sold separately.
Specifications
Product Type Roller Mop Head
Autoclavable Autoclavable
For Use With (Application) Disinfectants, Alcohol Floor, Walls, Ceilings
Material NovaLite
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.