Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Micronova™ SST™ Striped Safety Tape

Click to view available options
Color:
Black, White
Black, Yellow
Width (English):
1 in.
2 in.
3 in.
Description
For delineating and color coding specific equipment and areas
The Micronova SST™ Striped Safety Tape is a striped, pressure-sensitive tape with rubber adhesive that is ideal for marking fire equipment, exits, hazardous areas and first aid points within cleanrooms and controlled environments. The laminated vinyl is scuff resistant and holds up well to solvents, humidity and oil.
- Laminated vinyl film with rubber adhesive
- Cleanroom processed, pressure sensitive and scuff resistant
- Designed for delineating and color coding in cleanrooms and controlled environments
- Ideal for marking fire equipment, exits, hazardous areas and first aid points
- Available in black/yellow, red/white, and black/white stripes
- Holds up well to solvents, humidity and oil
- Wound on a 3-inch plastic core
Order Info
Available in 1, 2 or 3 in. widths.
Specifications
Specifications
Autoclavable | Not Autoclavable |
Adhesive Type | Rubber |
Color | Black, White |
Material | Vinyl |
Product Type | Cleanroom Tape |
Sterility | Non-sterile |
For Use With (Application) | Delineating and color-coding specific equipment and areas |
Temperature Range (English) | 60°F to 80°F |
Temperature Range (Metric) | 16°C to 27°C |
Dimensions (LxW) | 16.5 m x 50.8 mm (18 yd. x 2 in.) |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction