Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vileda Professional™ MicroOne™ Pocket Mop

Catalog No. p-8687000 Shop All Vileda Professional Products
Click to view available options
Color:
Red
This item is not returnable. View return policy
This item is not returnable. View return policy

MicroOne™ Mops offer microfiber cleaning performance in a disposable format, with a sturdy feel of a durable mop.

  • Microfiber cleaning performance in a disposable mop
  • Fast, stubborn dirt removal and spill pick-up
  • Double the absorbency of compatible mops
  • Improved fit onto the frame
  • Fits CombiSpeed™ Pro Frame in addition to other common industry frames
  • 4 Colors available
  • Low friction and superb ergonomics
  • Compliant with a wide range of cleaning solutions
  • Single use concept reduces the risk of cross-contamination

Specifications

Product Type Disposable Microfiber Pocket Mop
Color Red
Sterility Non-sterile
Autoclavable Not Autoclavable
Size 16 in.
Disposable Single-use
Quantity 25/Pk., 5 Pk./Cs.
For Use With (Equipment) CombiSpeed Frame (143572) Telescopic Handle (119966)
For Use With (Application) Bucket or pre-prepared Method
Material Microfiber
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.