Description
- For use with Agilent 5890, 6890, and 7890 GCs
- Sample pathways deactivated for ultimate inertness
Specifications
Specifications
| For Use With (Application) | FID |
| Includes | FID Adaptor (Large Bore), 1/4 in. Ferrule (Vespel/Graphite), 1/4 in. Nut (Stainless Steel), 1/16 in. Nut (Stainless Steel), 1/16 in. Ferrule (Vespel/Graphite) |
| For Use With (Equipment) | Packed Column FIDs Only |
| Functionality/Interaction Type | Sample pathways deactivated for ultimate inertness |
| Product Type | Micropacked Column FID Adaptor Kit |