Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Microphthalmia Transcription Factor (MiTF) Mouse anti-Human, Mouse, Rat, Clone: C5, Exalpha
SDP

Catalog No. 502101970
Encompass
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-210-1970 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-210-1970 Supplier Vector Laboratories Supplier No. X1405M
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Microphthalmia Transcription Factor (MiTF) Monoclonal antibody specifically detects Microphthalmia Transcription Factor (MiTF) in Human, Mouse, Rat samples. It is validated for Gel Shift, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot

Specifications

Antigen Microphthalmia Transcription Factor (MiTF)
Applications Gel Shift, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone C5
Concentration 0.5-2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.08% sodium azide; pH
Gene MITF
Gene Accession No. O75030, O88368, Q08874
Gene Alias BCC2; Bhlhe32; black eyed white; Bw; Class E basic helix-loop-helix protein 32; CMM8; Gsfbcc2; melanogenesis associated transcription factor; MI; microphtalmia-associated transcription factor; microphthalmia; microphthalmia-associated transcription factor; MITF; MITF-H; MITF-M; vit; Vitiligo; Wh; WS2; WS2A
Host Species Mouse
Immunogen Hybridoma produced by the fusion of splenocytes from RBF/DnJ mice immunized with an N-terminal fragment of human microphthalmia protein and mouse myeloma NS1 cells.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17342, 25094, 4286
Target Species Human, Mouse, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.