Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Micropipette Tips, Bulk
SDPSCI_ED

Catalog No. S8538036 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Volume (Metric):
10 μL
20 μL
200 μL
1000 μL
5 mL
10 mL
Color:
Clear Blue
Natural
Yellow
Graduated:
No
Yes
Model:
T1000UL-B
T1000UL-N
T10ML-N
T10UL-N
T200UL-N
T200UL-Y
T20UL-N
T5ML-N
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Color Graduated Model
S8538036 20 μL Natural No T20UL-N
S8538035 10 μL Natural No T10UL-N
S8538038 200 μL Natural No T200UL-N
S8538037 200 μL Yellow No T200UL-Y
S8538040 1000 μL Natural No T1000UL-N
S8538039 1000 μL Clear Blue Yes T1000UL-B
S8538041 5 mL Natural No T5ML-N
S8538042 10 mL Natural No T10ML-N
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. S8538036 Supplier United Scientific™ Supplier No. T20ULN

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

View Alternate Product

Micropipette Tips are disposable tips intended for use with most brands of pipettors.

  • The highly smooth, hydrophobic surface provides low liquid retention and increased accuracy and repeatability.
  • Inner radial rings enable an airtight grip on the tip cone.
  • Manufactured in a precision molding facility from 100% virgin polypropylene.
  • Free of DNase, RNase and pyrogen.

Specifications

Description Micropipette Tips, PP, 20 μL, Natural
Volume (Metric) 20 μL
Filtered Non-filtered
Sterility Non-sterile
Color Natural
Autoclavable Autoclavable
Model T20UL-N
Certifications/Compliance DNase, RNase and Pyrogen free
For Use With (Application) Pipetting
Format Bagged
Tip Style Standard
Packaging Bagged
Quantity 1000 Pk.
Disposable Single-use
For Use With (Equipment) Universal Fit
Graduated No
Material Polypropylene
Graduations No
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.