Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Micropipette Tips, Low Retention, Bulk
SDPSCI_ED

Catalog No. S8538031
Click to view available options
Volume (Metric):
10 μL
1000 μL
200 μL
300 μL
Tip Style:
Beveled, Low Retention
Extended Length, Low Retention
Low Retention
UltraFine, Low Retention

Low Retention Micropipette Tips are disposable tips intended for use with most brands of pipettors.

  • Deliver 10x lower retention levels with viscous samples, as compared to standard pipette tips.
  • Highly homogenous surface is free of defects, preventing non-specific binding.
  • Manufactured from ultra pure USP Class VI certified virgin polypropylene.
  • Free of DNase, RNase and pyrogen.
  • Natural color.
  • Bulk packaging.

Specifications

Description Low Retention Micropipette Tips
Volume (Metric) 200 μL
Filtered Non-filtered
Sterility Non-sterile
Color Natural
Autoclavable Autoclavable
Model UNP10003
Certifications/Compliance USP Class VI certified
For Use With (Application) Pipetting
Compatibility Universal
Format Bagged
Tip Style Beveled, Low Retention
Quantity 1000 Pk.
Product Type Micropipette Tips
Disposable Single-use
For Use With (Equipment) Universal Fit
Graduated No
Material Polypropylene
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.