Learn More
Description
- Black Eppendorf Microplates offer an excellent signal-to-noise ratio – for clear signals even with low-concentration samples.
- White Eppendorf Microplates are optimized for highest sensitivity in the detection of luminescence signals by maximizing reflection.
- Solid black and white assay plates are made of polypropylene resulting in high resistance to chemicals, mechanical stress, and temperature extremes.
- All plates are optimized for minimal autofluorescence and autoluminescence.
- Available in lot-certified PCR clean purity grade.
- Outstanding dimensional accuracy and high well-to well homogeneity for perfect automation suitability.
Specifications
Specifications
| Autoclavable | Autoclavable |
| Color | Gray |
| No. of Wells | 96 |
| Well Shape | Flat Bottom (F-well) |
| Max. RCF | 6000 x g |
| Purity or Quality Grade | PCR Clean (Non-sterile) |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.