Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eppendorf™ Assay/Reader Microplate

Catalog No. E951040102 Shop All Eppendorf North America Products
Change view
Click to view available options
Color:
Gray
White
Well Shape:
Conical Bottom (V-well)
Flat Bottom (F-well)
Round Bottom (U-well)
No. of Wells:
96
384
Well Color:
Black
White
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Well Shape No. of Wells Well Color
E951040102 White Round Bottom (U-well) 96 Black
05-414-244 White Flat Bottom (F-well) 96 -
05-414-243 Gray Flat Bottom (F-well) 96 -
E951040308 Gray Conical Bottom (V-well) 96 White
E951040145 Gray Round Bottom (U-well) 96 White
E951040260 White Conical Bottom (V-well) 96 Black
E951040503 Gray Conical Bottom (V-well) 384 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. E951040102 Supplier Eppendorf™ Supplier No. 951040102
Add to Cart
Add to Cart

Eppendorf Assay/Reader Microplates are designed for diverse microplate assays, optimizing absorbance, fluorescence, and chemiluminescence. With options for both top- and bottom-reading plate readers, they ensure clear signals even with low-concentration samples.

  • Black Eppendorf Microplates offer an excellent signal-to-noise ratio – for clear signals even with low-concentration samples.
  • White Eppendorf Microplates are optimized for highest sensitivity in the detection of luminescence signals by maximizing reflection.
  • Solid black and white assay plates are made of polypropylene resulting in high resistance to chemicals, mechanical stress, and temperature extremes.
  • All plates are optimized for minimal autofluorescence and autoluminescence.
  • Available in lot-certified PCR clean purity grade.
  • Outstanding dimensional accuracy and high well-to well homogeneity for perfect automation suitability.

Compliance

Compliant to SBS specifications

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance SBS
Format Standard
Array 8 x 12
Autoclavable Autoclavable
Color White
Product Type Microplate
Material Polypropylene
Volume (Metric) Well 360 μL
No. of Wells 96
Volume (Metric) Working 20 to 320 μL
Sterility Non-sterile
Well Shape Round Bottom (U-well)
For Use With (Application) Fluorescence detection
Max. RCF 6000 x g
Purity or Quality Grade PCR Clean (Non-sterile)
Well Color Black
Width (Metric) 12.77 cm
Height (Metric) 1.44 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.