Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sartorius Microsart™ AMP Coating Buffer

Catalog No. 14577842 Shop All Sartorius Products
Change view
Click to view available options
AMP Coating Buffer
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
14-577-842 AMP Coating Buffer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-577-842 Supplier Sartorius Supplier No. SMB952002

Increases recovery of Mycoplasma cells from filtration membrane. Sartorius Microsart™ AMP Coating Buffer is used in conjunction with Vivaspin™ units to improve sensitivity of detection system.

  • Used in combination with the Vivaspin™ units
  • High molecular weight components in physiological matrix
  • Increase recovery of Mycoplasma cells from the filtration membrane of the Vivaspin units
  • High detection sensitivity
  • Prevents unspecific binding of Mycoplasma cells

Compliance

EP 2.6.7

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material DNA extraction kit
For Use With (Application) DNA Extraction
For Use With (Equipment) Microstart AMP Quantitative detection kit, Vivaspin™ centrifugation columns
Quantity 2 x 20mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.