Learn More
Description
- Sensitive detection of Mycoplasma DNA
- Primer, probes designed against rRNA gene Mycoplasma species
- Sample type: Master cell banks, virus seed lots, virus harvests, final lot of biologicals produced by cell
- Detection limit: 10cfu/mL
- Sample volume: 200μl to 18mL
Applications
- Used in quality control analysis in microbiological and biopharmaceutical labs
Compliance
EP 2.6.7
Specifications
Specifications
| Content And Storage | Positive control, buffer, PCR grade water. 2° to 8°C |
| Detection Method | qPCR |
| No. of Tests | 25 tests |
| Product Type | Assay kit |
| Quantity | 25 tests |
| Target | 16s RNA of Mycoplasma |
| Sample Type | Lyophilized |
| Volume | 18 to 200μL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.