Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sartorius Microsart™ AMP Mycoplasma Quantitative Detection Kit

Catalog No. 14577840 Shop All Sartorius Products
Change view
Click to view available options
Quantity:
100 tests
25 tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-577-840 100 tests
14-577-839 25 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14-577-840 Supplier Sartorius Supplier No. SMB951002

missing translation for 'validMainframeMsg'

Rapid Real-Time PCR detection kit. Sartorius Microsart™ AMP Mycoplasma Quantitative Detection Kit is used for direct detection of Mollicutes in cell culture, media components and derived biologicals.

  • Sensitive detection of Mycoplasma DNA
  • Primer, probes designed against rRNA gene Mycoplasma species
  • Sample type: Master cell banks, virus seed lots, virus harvests, final lot of biologicals produced by cell
  • Detection limit: 10cfu/mL
  • Sample volume: 200μl to 18mL

Applications

  • Used in quality control analysis in microbiological and biopharmaceutical labs

Compliance

EP 2.6.7

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Positive control, buffer, PCR grade water. 2° to 8°C
Detection Method qPCR
No. of Tests 100 tests
Product Type Assay kit
Quantity 100 tests
Target 16s RNA of Mycoplasma
Sample Type Lyophilized
Volume 18 to 200μL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.