Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sartorius Microsart™ Research Mycoplasma Real-Time PCR Kit

Catalog No. 14577837 Shop All Sartorius Products
Change view
Click to view available options
Quantity:
100 tests
25 tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-577-837 25 tests
14-577-838 100 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14-577-837 Supplier Sartorius Supplier No. SMB951005

For fast and reliable screening of cell culture supernatants. Microsart™ Research Mycoplasma Real-Time PCR Kit is used for direct detection of Mollicutes in cell culture, media components, and derived biologicals.

  • Enables fast and reliable detection of Mycoplasma DNA in cell culture supernatants
  • Detects 16 S rRNA gene of at least 110 Mycoplasma species
  • Time: Three hours
  • Sample can be added directly to PCR reaction tube
  • TaqMan™ real-time qPCR based chemistry
  • Sample volume: 200μL

Applications:

  • Used with any Real-time PCR instrument which detects the dyes FAM, ROX
  • Used in quality control analysis in microbiological and biopharmaceutical labs

Compliance

EP 2.6.7

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2° to 8°C
Detection Method qPCR
No. of Tests 25 tests
Product Type Assay test
Quantity 25 tests
Target 16s rRNA of Mycoplasma
For Use With (Application) Microbial Detection
No. of Reactions 25 reactions
Sample Type Lyophilized
Volume 2μL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.