Learn More
Fisher Science Education™ Microscope Replacement Bulb
GREENER_CHOICESCI_ED
Catalog No. s72623 Shop All Fisherbrand Products
Click to view available options
For Use With (Equipment):
160 Series stereoscopic microscopes
Microscope Replacement Bulb for use with a variety of microscopes
  • Light bulb for use with 160 Series stereoscopic microscopes
  • 20 watt, 12 V
Specifications
Type Replacement microscope light bulb
Description G4 Pin base
For Use With (Equipment) 160 Series stereoscopic microscopes
Voltage 12 V
Warranty 5 years optical and mechanical, 1 year electrical from date of purchase
Wattage 20 w
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.