Learn More
United Scientific™ Microscope Slide Mailer
SDPSCI_ED
Catalog No. S15754 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Slide Capacity:
1
2
5
10
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Slide Capacity
S15754 2
S15743 1
S15939 5
S15859 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. S15754 Supplier United Scientific™ Supplier No. 62102
Add to Cart
Add to Cart
Allows for the safe storage and transport of microscope slides.
  • These white polypropylene slide mailers included hinged covers and molded-in dividers to securely hold 3 x 1 in. microscope slides.
  • Available in four sizes.
  • These white polypropylene slide mailers included hinged covers and molded-in dividers to securely hold 3 x 1 in. microscope slides.
  • Available in four sizes.
Specifications
Type Slide Mailer
Slide Capacity 2
For Use With (Equipment) Microscope Slides