Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Microscope Slide-Making Kit
SCI_ED

Catalog No. s06972 Shop All Aldon Products
Click to view available options
:
Microscope Slide Making Kit
WARNING: This product can expose you to chemicals including Ethylbenzene, which is known to the State of California to cause cancer.
WARNING: This product can expose you to chemicals including Ethylbenzene, which is known to the State of California to cause cancer.

Includes

  • 25 mL synthetic balsam
  • 72 microscope slides
  • 100 cover slips
  • One set of forceps

Make your own slides in your lab or classroom.

Basic slide-making kit will get you started mounting your own slides.

Specifications

Certifications/Compliance 49 CFR 173.4
Product Type Microscope Slide Making Kit
Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) This basic microscope slide-making kit will get you started mounting your own slides, for lab or classroom
Includes 25 mL synthetic balsam, 1 microscope slide, 1 coverslips, 1 forceps
Quantity 1
Dimensions (L x W x H) 4.25 x 4.625 x 5.5 in.
Class Size 75 Students
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.