Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Simport™ Scientific Microscreen™ I Small Biopsy Processing/Embedding Cassettes
Click to view available options
Color:
Orange
Peach
Yellow
Description
- Microscreen™ cassette is made of a strong nylon mesh
- Designed to handle small, fragile biopsies
- Nylon mesh has 1,500 square openings (300 μm each)
- 8 x 11 mm slots in base and lid to help cassette sink rapidly
- Sturdy nylon mesh improves fluid exchange by reducing surface tension of reagents
- Accelerates fixation/preparation of tissue samples
- Eliminates the need for pads, sponges, nylon or lens paper bags
- Reduces the number of incidents involving non-fixed or insufficiently fixed tissues
- Inner chamber design for greater biopsy protection
- Large front tab for easier lid opening
- Hinge design configuration allows for multiple opening and closing
Specifications
Specifications
Writing Face Angle | 45° |
Color | Orange |
Material | Acetal |
Product Type | Cassette |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction