Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Supertek™ Microslide™ Drugs and Nutrition

Shop All Supertek Scientific Products


Click to view available options
Slide Type:
Photomicrograph Slide
Description
- Image titles on Microslide:
- 1. Smoking-Lung (Normal 130x , Abnormal 50x)
- 2. Glue Sniffing - Liver (Normal and Abnormal 100x)
- 3. Alcohol - Esophagus (Normal and Abnormal (20x)
- 4. Aspirin - Stomach (Normal and Abnormal 100x)
- 5. Vitamin D Deficiency - Bones (Normal and Abnormal (40x)
- 6. Iron Deficiency - Bones (Normal and Abnormal (1300x)
- 7. Iodine Deficiency - Thyroid (Normal 40x, Abnormal 100x)
- 8 Vitamin B1 Deficiency - Heart Muscle ( Normal 100x, Abnormal 250x)
- Comes with a Lesson Plan with Pocket to store Microslide.
- Reproducible Worksheet and Storage Box with a purchase of 15.
- Grades 6-12
Specifications
Specifications
Classification | Human Physiology |
Material | Paper and Film |
Type | Microslide |
Grade | 6 to 12 |
Includes | Microslide and Lesson Plan |
Slide Type | Photomicrograph Slide |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction