Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Supertek™ Microslide™ How a One - Celled Animal Divides

Shop All Supertek Scientific Products


Click to view available options
Slide Type:
Photomicrograph Slide
Description
- Image titles on Microslide:
- 1. Paramecia - Fresh (10x)
- 2. Paramecium - Fresh -(100x)
- 3. Paramecium - Stained (100x)
- 4. Division - Stage 1 - Stained (100x)
- 5. Stage 2 - Stained - (100x)
- 6. Stage 3 - Stained - (100x)
- 7. Stage 4 - Stained - (100x)
- 8. Paramecium - All Stages - Stained (10x)
- Comes with an Extensive Lesson Plan with a Pocket to store the Microscope.
- Reproducible Worksheet and Storage Box with a purchase of 15.
- Grades 9-12
Specifications
Specifications
Classification | Animal Development |
Material | Paper and Film |
Type | Microslide |
Grade | 9 to 12 |
Includes | Microslide and Lesson Plan |
Slide Type | Photomicrograph Slide |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction