Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Supertek™ Microslide™ How Rocks are Formed

Shop All Supertek Scientific Products


Click to view available options
Grade:
1 to 5
Description
- Image titles on Microslide:
- 1. Granite (50x)
- 2. Basalt (50x)
- 3. Pyroxenite (50x)
- 4. Red Sandstone (50x)
- 5. Shell Limestone (50x)
- 6. Quartzite (50x)
- 7. Mica Schist (50x)
- 8. Marble (50x)
- Comes with a Lesson Plan with Pocket to store Microslide.
- Reproducible Worksheet and Storage Box with a purchase of 15.
- Grades 1-5
Specifications
Specifications
Classification | Geology |
Material | Paper and Film |
Type | Microslide |
Grade | 1 to 5 |
Includes | Microslide and Lesson Plan |
Slide Type | Photomicrograph Slide |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction