Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Supertek™ Microslide™ Immunity

Shop All Supertek Scientific Products


Click to view available options
Slide Type:
Photomicrograph Slide
Description
- Image titles on Microslide:
- 1. Blood Phagocyte (900x)/Phagocytosis (2500x)
- 2. Monocyte (3000x)/Macrophage (4000x)
- 3. Bone Marrow (400x)/Lymph Gland (40x)
- 4. Thymus Gland (175x)
- 5. Normal T-Cells (2000x)/HIV budding from T-Cell (2700x)
- 6. Plasma Cell (37000x)
- 7. Normal Red Blood Cells (3000x) -Agglutinated Cells (100x)
- 8. Eosinophil (1400x)/Basophil (1400x)
- Comes with an Extensive Lesson Plan with a Pocket to store the Microscope.
- Reproducible Worksheet and Storage Box with a purchase of 15.
- Grades 6-12
Specifications
Specifications
Classification | Pathogens and Diseases |
Material | Paper and Film |
Type | Microslide |
Grade | 6 to 12 |
Includes | Microslide and Lesson Plan |
Slide Type | Photomicrograph Slide |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction