Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Supertek™ Microslide™ Life Functions of Paramecia

Shop All Supertek Scientific Products


Click to view available options
Slide Type:
Photomicrograph Slide
Description
- Image titles on Microslide:
- 1. Paramecium with Bacteria (100x)/Paramecia with Air Bubble (40x)
- 2. Ingestion (200x)/Digestion (100x)
- 3. Paramecium Dividing (100x)
- 4. Paramecium (200x)
- 5. Contractile Vacuoles (200x)
- 6. Paramecium Nuclei (200x)/Conjugation (100x)
- 7. Paramecium Bending (100x)
- 8. Trichocysts (100x)
- Lesson Plan with Pocket to store Microslide
- Reproducible Worksheet and Storage Box with a purchase of 15
- Grades 9-12
Specifications
Specifications
Classification | Protists |
Material | Paper and Film |
Type | Microslide |
Grade | 9 to 12 |
Includes | Microslide and Lesson Plan |
Slide Type | Photomicrograph Slide |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction