Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Supertek™ Microslide™ Life in the Desert

Shop All Supertek Scientific Products


Click to view available options
Slide Type:
Photomicrograph Slide
Description
- Image titles on Microslide:
- 1.Saguaro Cactus
- 2. Gila Woodpecker in Saguaro Nest
- 3. Field of Wildflowers
- 4. Kangaroo Rat
- 5. Gila Monster
- 6. Jack Rabbit
- 7. Tarantula
- 8. Scorpion
- Comes with an Extensive Lesson Plan with a Pocket to store the Microscope.
- Reproducible Worksheet and Storage Box with a purchase of 15.
- Grades 6-12
Specifications
Specifications
Classification | Biological Adaptations |
Material | Paper and Film |
Type | Microslide |
Grade | 6 to 12 |
Includes | Microslide and Lesson Plan |
Slide Type | Photomicrograph Slide |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction