Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Supertek™ Microslide™ Taxonomy - Classification and Keys

Shop All Supertek Scientific Products


Click to view available options
Slide Type:
Photomicrograph Slide
Description
- Image titles on Microslide:
- 1. Bacteria in Pairs - (1200x)
- 2. A Spore Producer - Stained - (1000x)
- 3. Bacteria in Chains - Stained - (1000x)
- 4. Rod Shaped Bacteria - Stained - (2000x)
- 5. Another Spore Producer - Stained - (1000x)
- 6. Boils and Abscess Bacteria - Stained - (1000x)
- 7. Syphilis Bacterium - Electron Micrograph - (30,000x)
- 8. Sauerkraut Bacteria - Stained - (2000x)
- Comes with an Extensive Lesson Plan with a Pocket to store the Microscope.
- Reproducible Worksheet and Storage Box with a purchase of 15.
- Grades 6-12
Specifications
Specifications
Classification | Microbiology |
Material | Paper and Film |
Type | Microslide |
Grade | 6 to 12 |
Includes | Microslide and Lesson Plan |
Slide Type | Photomicrograph Slide |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction