Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Supertek™ Microslide™ The Endocrine System

Shop All Supertek Scientific Products


Click to view available options
Grade:
9 to 12
Description
- Image titles on Microslide:
- 1. The Pancreas (25x)
- 2. Islands of Langerhans (190x)
- 3. Thyroid Gland (420x)
- 4. Parathyroid Gland (45x)
- 5. Adrenal Gland (7x)
- 6. Adrenal Cortex (180x)
- 7. Pituitary Gland (5x)
- 8. Anterior Pituitary (90x)
- Comes with an Extensive Lesson Plan with a Pocket to store the Microscope.
- Reproducible Worksheet and Storage Box with a purchase of 15.
- Grades 9-12
Specifications
Specifications
Classification | Human Physiology |
Material | Paper and Film |
Type | Microslide |
Grade | 9 to 12 |
Includes | Microslide and Lesson Plan |
Slide Type | Photomicrograph Slide |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction