Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Supertek™ Microslide™ The Frog

Shop All Supertek Scientific Products


Click to view available options
Slide Type:
Photomicrograph Slide
Description
- Image titles on Microslide:
- 1.Frog Skin (160X
- 2. Frog Gills
- 3. Frog Lungs l.s. stained (1X)
- 4. Frog Blood (400X) / Human Blood (900X)
- 5. Frog Heart left-w.m. (2X) / right- l.s. (5X) L
- 6. Frog Intestine -X.S. (20X)
- 7. Frog Cloaca (2X)
- 8. Frog Brain w.m. (2X)
- Comes with an Extensive Lesson Plan with a Pocket to store the Microslide.
- Reproducible Worksheet and Storage Box with a purchase of 15.
- Grades 9-12
Specifications
Specifications
Classification | Diversity of Animal Life |
Material | Paper and Film |
Type | Microslide |
Grade | 9 to 12 |
Includes | Microslide and Lesson Plan |
Slide Type | Photomicrograph Slide |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction