Learn More
Description
- Image titles on Microslide:
- 1. Lilac Leaf - x.s. Stained (60x)
- 2. Working layers of the Leaf
- 3. Upper Epidermis and Palisades
- 4. Chloroplast - Electron Micrograph (30,000x)
- 5. Epidermis - Surface View - Stained (150x)
- 6. Stomate - Surface View - Stained (430x)
- 7. Air Space and Spongy Layer x.s. - Stained (480x)
- 8. Abscissions Layer - I.s. - Stained (12x)
- Lesson Plan with Pocket to store Microslide
- Reproducible Worksheet and Storage Box with a purchase of 15
- Grades 6-12
Specifications
Specifications
| Classification | Botany |
| Material | Paper and Film |
| Type | Microslide |
| Grade | 6 to 12 |
| Includes | Microslide and Lesson Plan |
| Slide Type | Photomicrograph Slide |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

