Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Supertek™ Microslide™ The Microscope - A Tool for Learning

Shop All Supertek Scientific Products


Click to view available options
Slide Type:
Photomicrograph Slide
Description
- Image titles on Microslide:
- 1. The Letter E (100x)
- 2. Cheek Cells (100x and 400x)
- 3. Cheek Cells (900x)
- 4. Onion Skin Cells (110x)
- 5. Hydra (50x) and (175x)
- 6. Paramecium (200x)
- 7. Paramecium-Interference Microscope (200x)
- 8. Paramecium (1500x)
- Comes with an Extensive Lesson Plan with a Pocket to store the Microscope.
- Reproducible Worksheet and Storage Box with a purchase of 15.
- Grades 6-8
Specifications
Specifications
Classification | Microscopy |
Material | Paper and Film |
Type | Microslide |
Grade | 6 to 8 |
Includes | Microslide and Lesson Plan |
Slide Type | Photomicrograph Slide |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction