Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Supertek™ Microslide™ The Moon

Shop All Supertek Scientific Products


Click to view available options
Slide Type:
Photomicrograph Slide
Description
- Image titles on Microslide:
- 1. The (almost) Full Moon
- 2. Craters and More Craters
- 3. Lunar Rock and Roll
- 4. Mountains and Plains
- 5. Crater Copernicus
- 6. Lunar Sightseeing
- 7. The Tides (the Bay of Fundy)
- 8. A Lunar Eclipse
- Lesson Plan with Pocket to store Microslide
- Reproducible Worksheet and Storage Box with a purchase of 15
- Grades 6-8
Specifications
Specifications
Classification | Astronomy |
Material | Paper and Film |
Type | Microslide |
Grade | 6 to 8 |
Includes | Microslide and Lesson Plan |
Slide Type | Photomicrograph Slide |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction