Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Supertek™ Microslide™ The Root of a Flowering Plant

Shop All Supertek Scientific Products


Click to view available options
Slide Type:
Photomicrograph Slide
Description
- Image titles on Microslide:
- 1. Buttercup Root - x.s. - Stained (40x)
- 2. Central Cylinder - x.s. - Stained (220x)
- 3. Epidermis and Cortes - x.s. Stained (75x)
- 4. Root Cap and Zone of Growth - I.s. -Stained (34x)
- 5. Zone of Elongation - I.s. - Stained (34x)
- 6. Zone of Maturation - I.s. - Stained (45x)
- 7. Root Tip - Root Hairs - w.m. - Stained (45x)
- 8. Epidermal Cell - Root Hair - I.s. - Stained (260x)
- Lesson Plan with Pocket to store Microslide
- Reproducible Worksheet and Storage Box with a purchase of 15
- Grades 6-12
Specifications
Specifications
Classification | Botany |
Material | Paper and Film |
Type | Microslide |
Grade | 6 to 8 |
Includes | Microslide and Lesson Plan |
Slide Type | Photomicrograph Slide |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction