Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Supertek™ Microslide™ The Stem of a Flowering Plant

Shop All Supertek Scientific Products


Click to view available options
Slide Type:
Photomicrograph Slide
Description
- Image titles on Microslide:
- 1. Corn Stem - x.s. - Stained (8x)
- 2. Fibrovascular Bundle (corn) x.s. - Stained (170x)
- 3. Red Clover Stem - x.s. Stained (34x)
- 4. Basswood Stem (1 year Old) x.s. - Stained (18x)
- 5. Basswood Stems (2 and 3 years old) - x.s. Stained (18x)
- 6. An Annual Ring - x.s. Stained (76x)
- 7. Coleus Bud - I.s. Stained (55x)
- 8. Lenticel in Elderberry Stem - x.s. - Stained (55x)
- Lesson Plan with Pocket to store Microslide
- Reproducible Worksheet and Storage Box with a purchase of 15
- Grades 6-12
Specifications
Specifications
Classification | Botany |
Material | Paper and Film |
Type | Microslide |
Grade | 6 to 12 |
Includes | Microslide and Lesson Plan |
Slide Type | Photomicrograph Slide |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction