Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Microsomal Glutathione S-transferase 1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP30295320UL
Description
Microsomal Glutathione S-transferase 1 Polyclonal specifically detects Microsomal Glutathione S-transferase 1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
Microsomal Glutathione S-transferase 1 | |
Polyclonal | |
PBS with 50% glycerol, pH7.3. | |
Rabbit | |
Affinity purified | |
RUO | |
4257 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Immunofluorescence | |
Unconjugated | |
glutathione S-transferase 12, GST12EC 2.5.1.18, MGC14525, MGST, MGST-I, microsomal glutathione S-transferase 1, Microsomal GST-1, Microsomal GST-I | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human Microsomal Glutathione S-transferase 1 (NP_001247440.1). | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction