Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ MicroTest™ Flocked Swab Kits
Click to view available options
Tip Type:
Microtip
Traditional
Traditional and Microtipped
Product Type:
Flocked Swab Kit
Universal Kit
Includes:
M6 Kit w/Micro-Tipped Flocked Swab
M6 Kit w/Traditional-Tipped Flocked Swab
Description
Maintain organism viability and counts for verification studies, quality control procedures, and repeat testing of viruses chlamydiae, mycoplasma and ureaplasma with quality media, proven collection and transport tools, and safe, reliable packaging with Thermo Scientific™ MicroTest™ Flocked Swab Kits.
Designed for the collection, transportation, maintenance and long-term storage of viral specimens, chlamydiae, mycoplasma and ureaplasma, Thermo Scientific MicroTest products are top quality. From formulation to packaging, MicroTest products maintain the viability of organisms through freeze-thaw cycles while inhibiting antimicrobial contaminants and promoting safety.
MicroTest™ M4RT™ contains gelatin, gentamicin, and amphotericin B for the transport of viruses and Chlamydiae.
MicroTest™ M6™ contains gelatin, vancomycin, amphotericin B, and colistin for the transport of viruses, Chlamydiae, Ureaplasmas, and Mycoplasmas.
Microtest Flocked Swab Kits come with 1.5mL or 3mL of liquid medium in a 15mL conical tube with glass beads, an envelope with one or two nylon-flocked swabs, and a 6 x 9 in. zip-seal bag.
Keep your lab moving in the right direction with a dependable set of solutions for transport, holding, maintenance and long-term culture storage with the MicroTest family of collection and transport media.
Trusted reliable results
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
MicroTest™ M4RT™ contains gelatin, gentamicin, and amphotericin B for the transport of viruses and Chlamydiae.
MicroTest™ M6™ contains gelatin, vancomycin, amphotericin B, and colistin for the transport of viruses, Chlamydiae, Ureaplasmas, and Mycoplasmas.
Microtest Flocked Swab Kits come with 1.5mL or 3mL of liquid medium in a 15mL conical tube with glass beads, an envelope with one or two nylon-flocked swabs, and a 6 x 9 in. zip-seal bag.
Keep your lab moving in the right direction with a dependable set of solutions for transport, holding, maintenance and long-term culture storage with the MicroTest family of collection and transport media.
Trusted reliable results
- Stringent quality control, including cellular toxicity on each lot number
- Patented formulations with stabilizing proteins, buffers, and specific antibiotics
- Provides compatible transport for cell culture, enzyme immunoassays, and molecular diagnostic tests
- Maintains organism viability and counts for better, more reliable results during validation/verification studies, quality control procedures, and repeat testing
- 1.5mL and 3.0mL fill volumes allow multiple tests from one specimen
- Individual transport media tubes or complete kits offer flexibility and versatility
- Tough, break-resistant tubes with secure, color-coded screw caps prevent leaking and exposure to infectious agents
- Kits include leak-resistant polypropylene bag for safe sample transport
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.

Specifications
Specifications
Product Type | Flocked Swab Kit |
Format | Tube |
Media Type | M6 |
Swab Configuration | Single Applicator |
Closure Color | Red |
Compatibility | Culture, Shell vial, Freeze-thaw cycles, Culture maintenance, ELISA, PCR, DNA probe technology |
Fill Volume | 1.5 mL |
For Use With (Application) | Viruses: Chlamydiae, Ureaplasmas, and Mycoplasmas |
Includes | M6 Kit w/Micro-Tipped Flocked Swab |
Quantity | 100 Kits/Pk. |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction