Learn More
Stoelting™ Microvascular Clip
SDP
Stoelting™ Microvascular Clips are the most gentle of available Stoelting&Trade; clips. They apply light pressure to make sure no damage is done to the walls of the occluded vessel.
$262.00 - $304.50
Specifications
For Use With (Application) Neuroscience
View More Specs
Products 3
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
10-001-363
SDP
View Documents
Stoelting™
5212000
8 mm
Each for $304.50
Only null left
Add to Cart
10-001-364
SDP
View Documents
Stoelting™
5212002
11 mm
Each for $297.00
Only null left
Add to Cart
10-001-365
SDP
View Documents
Stoelting™
5212004
14 mm
Each for $262.00
Only null left
Add to Cart
Description
Description
Specifications
Specifications
Neuroscience
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.