Learn More
Argos Technologies™ Micryo Label Rolls
SCI_ED
Made from chemically inert polyester
Supplier:  Argos Technologies™ L1287/EMD
 View more versions of this product
Catalog No. S91001C
Description
Description
  • Unique adhesive formulated to adhere to laboratory glass and plasticware
  • Withstand conventional freezing (-80°C) and can also be used for most cryogenic storage applications (vapor and liquid phase nitrogen storage at (-196°C)
  • No need for separate labeling solutions for cryo-preservation units or ultra low freezers
  • Can even be used in incubators
Specifications
Specifications
Red
Polyester
1.5 to 2 mL tube, can even be used in incubators
1000/Roll
3.3 x 1.3 cm
Label
Micryo Strip
-196°C
1.29 x 0.5 in.
-320.8°F
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.