Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MICS1 Monoclonal Antibody (42), Invitrogen™
GREENER_CHOICE

Catalog No. PIMA543048
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA543048 100 μL
PIMA543482 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIMA543048 Supplier Invitrogen™ Supplier No. MA543048
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Specificity: Dermatophagoides pteronyssinus GHITM/MICS1

Required for the mitochondrial tubular network and cristae organization. Involved in apoptotic release of cytochrome c.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MICS1
Applications ELISA
Classification Monoclonal
Clone 42
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Alias 1010001P14Rik; C77840; dermal papilla-derived protein 2; DERP2; DKFZp566C0746; FLJ26584; GHITM; growth hormone inducible transmembrane protein; growth hormone-inducible transmembrane protein; growth hormone-inducible transmembrane protein-like protein; HSPC282; MICS1; mitochondrial morphology and cristae structure 1; My021; PTD010; similar to Dermal papilla derived protein 2; Tmbim5; transmembrane BAX inhibitor motif containing 5; transmembrane BAX inhibitor motif-containing protein 5; UNQ244/PRO281
Host Species Mouse
Immunogen Recombinant Dermatophagoides pteronyssinus GHITM/MICS1 Protein.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Arthropod
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.