Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MICS1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553426
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553426 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553426 Supplier Invitrogen™ Supplier No. PA553426
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DTQKVIKRAE VSPMYGVQKY DPINSMLSIY MDTLNIFMRV ATMLATGGNR KK Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 87%.

Required for the mitochondrial tubular network and cristae organization. Involved in apoptotic release of cytochrome c.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MICS1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GHITM
Gene Accession No. Q9H3K2
Gene Alias 1010001P14Rik; C77840; dermal papilla-derived protein 2; DERP2; DKFZp566C0746; FLJ26584; GHITM; growth hormone inducible transmembrane protein; growth hormone-inducible transmembrane protein; growth hormone-inducible transmembrane protein-like protein; HSPC282; MICS1; mitochondrial morphology and cristae structure 1; My021; PTD010; similar to Dermal papilla derived protein 2; Tmbim5; transmembrane BAX inhibitor motif containing 5; transmembrane BAX inhibitor motif-containing protein 5; UNQ244/PRO281
Gene Symbols GHITM
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MICS1. Recombinant protein control fragment (Product #RP-91271).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27069
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.