Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MICU1 Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA5142722
This item is not returnable.
View return policy
Description
Additional Information: This antibody is expected to recognize reported isoforms 1 and 2 (NP_006068.2; NP_001182447.1). This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:32,000.
MICU1 is a key regulator of mitochondrial calcium uptake required for calcium entry into mitochondrion. It may act as a calcium sensor via its EF-hand domains, gating the activity of a calcium channel partner. MICU1 induces T-helper 1-mediated autoreactivity, which is accompanied by the release of IFNG.
Specifications
| MICU1 | |
| Polyclonal | |
| Unconjugated | |
| MICU1 | |
| ara CALC; atopy-related autoantigen CALC; C730016L05Rik; CALC; calcium binding atopy-related autoantigen 1; calcium uptake protein 1, mitochondrial; calcium-binding atopy-like autoantigen 1; calcium-binding atopy-related autoantigen 1; Calcium-binding atopy-related autoantigen 1 homolog; calcium-binding atopy-related autoantigen-like protein; cbara1; DKFZp564C246; EFHA3; FLJ12684; Hom s 4; MICU1; mitochondrial calcium uptake 1; MPXPS; si:ch211-199i24.4; zgc:162148 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 10367 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q9BPX6 | |
| MICU1 | |
| Peptide with sequence C-EYENRIRAYSTPDK (aa 113-126). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction