Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MICU1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583371
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583371 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583371 Supplier Invitrogen™ Supplier No. PA583371
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: IFYTLGECGL ISFSDYIFLT TVLSTPQRNF EIAFKMFDLN GDGEVDMEEF EQVQSIIRSQ TSMGMRHRDR PTTGNTLKSG LCSALTTY Highest antigen sequence indentity to the following orthologs: Mouse - 99%, Rat - 100%.

MICU1 is a key regulator of mitochondrial calcium uptake required for calcium entry into mitochondrion. It may act as a calcium sensor via its EF-hand domains, gating the activity of a calcium channel partner. MICU1 induces T-helper 1-mediated autoreactivity, which is accompanied by the release of IFNG.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MICU1
Applications Western Blot, Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MICU1
Gene Accession No. Q9BPX6
Gene Alias ara CALC; atopy-related autoantigen CALC; C730016L05Rik; CALC; calcium binding atopy-related autoantigen 1; calcium uptake protein 1, mitochondrial; calcium-binding atopy-like autoantigen 1; calcium-binding atopy-related autoantigen 1; Calcium-binding atopy-related autoantigen 1 homolog; calcium-binding atopy-related autoantigen-like protein; cbara1; DKFZp564C246; EFHA3; FLJ12684; Hom s 4; MICU1; mitochondrial calcium uptake 1; MPXPS; si:ch211-199i24.4; zgc:162148
Gene Symbols MICU1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MICU1. Recombinant protein control fragment (Product #RP-104672).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10367
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.