Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MICU1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109325
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109325 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109325 Supplier Invitrogen™ Supplier No. PA5109325
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Control: Mouse kidney tissue, rat brain tissue, Hela.

MICU1 is a key regulator of mitochondrial calcium uptake required for calcium entry into mitochondrion. It may act as a calcium sensor via its EF-hand domains, gating the activity of a calcium channel partner. MICU1 induces T-helper 1-mediated autoreactivity, which is accompanied by the release of IFNG.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MICU1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4
Gene MICU1
Gene Accession No. Q9BPX6
Gene Alias ara CALC; atopy-related autoantigen CALC; C730016L05Rik; CALC; calcium binding atopy-related autoantigen 1; calcium uptake protein 1, mitochondrial; calcium-binding atopy-like autoantigen 1; calcium-binding atopy-related autoantigen 1; Calcium-binding atopy-related autoantigen 1 homolog; calcium-binding atopy-related autoantigen-like protein; cbara1; DKFZp564C246; EFHA3; FLJ12684; Hom s 4; MICU1; mitochondrial calcium uptake 1; MPXPS; si:ch211-199i24.4; zgc:162148
Gene Symbols MICU1
Host Species Rabbit
Immunogen Recombinant protein within Human CBARA1 aa 14-154.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10367
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles, store in dark
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.