Learn More
MICU3 Polyclonal Antibody, Invitrogen™
Description
Sequence homology: Cow: 91%; Dog: 75%; Human: 100%; Pig: 91%; Rabbit: 92%; Rat: 100%
Specifications
Specifications
| Antigen | MICU3 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide; pH 7.4 |
| Gene | MICU3 |
| Gene Accession No. | Q86XE3 |
| Gene Alias | calcium uptake protein 3, mitochondrial; EF hand domain family A2; EF hand domain family, member A2; EFHA2; EF-hand domain family, member A2; EF-hand domain-containing family member A2; MICU3; mitochondrial calcium uptake family member 3; mitochondrial calcium uptake family, member 3; mitochondrial uptake family, member 3 |
| Gene Symbols | MICU3 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.