Learn More
K1 Series Mid-Sole Ice Cleat
The K1 Series Mid-Sole ice cleat is the one-size-fits-all winter traction solution. The ice-penetrating tungsten carbide spikes maximize grip while the rotatable design provides seamless transition when traction is not required.
Supplier: K1 Series V9770350O/S
This item is not returnable.
View return policy
Description
- Available in Original, Low-Profile, and High-Profile styles to provide near-universal coverage of all footwear
- Strategically placed ice-penetrating tungsten carbide spikes provide maximum traction on ice and snow
- Easy to use design rotates to top of foot to allow driving, climbing, and indoor use
- Black or hi-viz adjustable strap delivers a perfect fit regardless of shoe size
- Specially formulated rubber pad provides maximum comfort and durability even in extreme cold weather conditions
Specifications
Elastic | |
Outdoor Surfaces, Ice Surfaces | |
Elastic | |
Reusable | |
Yes |
Black | |
Universal | |
High Profile Tungston Carbide Molded in | |
Unisex |