Learn More
K1 Series Mid-Sole Ice Cleat
Description
- Available in Original, Low-Profile, and High-Profile styles to provide near-universal coverage of all footwear
- Strategically placed ice-penetrating tungsten carbide spikes provide maximum traction on ice and snow
- Easy to use design rotates to top of foot to allow driving, climbing, and indoor use
- Black or hi-viz adjustable strap delivers a perfect fit regardless of shoe size
- Specially formulated rubber pad provides maximum comfort and durability even in extreme cold weather conditions
Specifications
Specifications
| Closure Type | Elastic |
| Color | Black |
| For Use With (Application) | Outdoor Surfaces, Ice Surfaces |
| Size | Universal |
| Strap Material | Elastic |
| Style | Original Profile Tungston Carbide Molded in |
| Disposable | Reusable |
| Gender | Unisex |
| Waterproof | Yes |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.