Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MID1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA599203
Description
Antibody detects endogenous levels of total TRI18.
Midline-1 (Tripartite motif-containing protein 18, Putative transcription factor XPRF, RING finger protein 59) is a 667 amino acid protein encoded by the human gene MID1. Midline-1 belongs to the TRIM/RBCC family and contains two B box-type zinc fingers, one B30.2/SPRY domain, one COS domain, one fibronectin type-III domain and one RING-type zinc finger. Midline-1 is believed to have E3 ubiquitin ligase activity which targets the catalytic subunit of protein phosphatase 2 for degradation. It is a cytoplasmic protein found as a homodimer or heterodimer with Midline-2. It also interacts with IGBP1 (Lymphocyte signaling protein A4). Defects in MID1 are the cause of Opitz syndrome type I (OS-I). OS-I is an X-linked recessive disorder characterized by hypertelorism, genital-urinary defects such as hypospadias in males and splayed labia in females, lip-palate-laryngotracheal clefts, imperforate anus, developmental delay and congenital heart defects.
Specifications
MID1 | |
Polyclonal | |
Unconjugated | |
MID1 | |
61B3-R; BBBG1; DXHXS1141; E3 ubiquitin-protein ligase Midline-1; Finger on X and Y (in rat only on X); Fxy; GBBB1; Mid1; Midin; midline 1; midline 1 (Opitz/BBB syndrome); midline 1 RING finger protein; Midline1; midline-1; OGS1; Opitz/BBB syndrome; OS; OSX; Putative transcription factor XPRF; RING finger protein 59; RING finger protein Midline-1; RING-type E3 ubiquitin transferase Midline-1; RNF59; TRIM18; tripartite motif protein TRIM18; tripartite motif-containing protein 18; XPRF; zinc finger on X and Y, mouse, homolog of; ZNFXY | |
Rabbit | |
Affinity chromatography | |
RUO | |
17318, 4281, 54252 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
O15344, O70583, P82458 | |
MID1 | |
A synthesized peptide derived from human MID1(Accession O15344), corresponding to amino acid residues I83-T133. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction