Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MID1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.50 - $499.50
Specifications
Antigen | MID1 |
---|---|
Applications | Western Blot, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
MID1 Polyclonal specifically detects MID1 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
MID1 | |
Polyclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
EC 6.3.2.-, FLJ57031, FLJ58683, FLJ76288, FXYGBBB1, Midin, midline 1 (Opitz/BBB syndrome), RNF59MIDIN, TRIM18mouse, homolog of | |
Recombinant fusion protein containing a sequence corresponding to amino acids 478-667 of human MID1 (NP_000372.1). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Western Blot, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Lipid and Metabolism, Zinc Finger | |
PBS with 50% glycerol, pH7.3. | |
4281 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title