Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MID1IP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA575714
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
PIPA575714 100 μL
1 options

Catalog No. PIPA575714

Supplier: Invitrogen™ PA575714

Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

Plays a role in the regulation of lipogenesis in liver. Up-regulates ACACA enzyme activity. Required for efficient lipid biosynthesis, including triacylglycerol, diacylglycerol and phospholipid. Involved in stabilization of microtubules.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MID1IP1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene MID1IP1
Gene Accession No. Q6P7D5, Q9CQ20, Q9NPA3
Gene Alias 3110038L01Rik; FLJ10386; G12-like; gastrulation specific G12 homolog; gastrulation-specific G12-like protein; MID1 interacting G12-like protein; MID1 interacting protein 1; MID1 interacting protein 1 (gastrulation specific G12 homolog); Mid1 interacting protein 1 (gastrulation specific G12-like (zebrafish)); MID1 interacting protein 1 (gastrulation specific G12-like); mid1-interacting G12-like protein; Mid1-interacting protein 1; MID1IP1; mid1ip1 {ECO:0000250; Mig12; Protein STRAIT11499; Protein STRAIT11499 homolog; S14R; slap; spot 14-like androgen-inducible protein; spot 14-R; spot 14-related protein; STRAIT11499; THRSPL; UniProtKB:Q9CQ20}
Gene Symbols MID1IP1
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues in Human MID1IP1.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 404280, 58526, 68041
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.