Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MID1IP1 Recombinant Rabbit Monoclonal Antibody (079)
GREENER_CHOICE

Catalog No. PIMA530676
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA530676 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA530676 Supplier Invitrogen™ Supplier No. MA530676
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. This antibody has specificity for Human MID1IP1.

Plays a role in the regulation of lipogenesis in liver. Up-regulates ACACA enzyme activity. Required for efficient lipid biosynthesis, including triacylglycerol, diacylglycerol and phospholipid. Involved in stabilization of microtubules.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MID1IP1
Applications ELISA
Classification Recombinant Monoclonal
Clone 079
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene MID1IP1
Gene Accession No. Q9NPA3
Gene Alias 3110038L01Rik; FLJ10386; G12-like; gastrulation specific G12 homolog; gastrulation-specific G12-like protein; MID1 interacting G12-like protein; MID1 interacting protein 1; MID1 interacting protein 1 (gastrulation specific G12 homolog); Mid1 interacting protein 1 (gastrulation specific G12-like (zebrafish)); MID1 interacting protein 1 (gastrulation specific G12-like); mid1-interacting G12-like protein; Mid1-interacting protein 1; MID1IP1; mid1ip1 {ECO:0000250; Mig12; Protein STRAIT11499; Protein STRAIT11499 homolog; S14R; slap; spot 14-like androgen-inducible protein; spot 14-R; spot 14-related protein; STRAIT11499; THRSPL; UniProtKB:Q9CQ20}
Gene Symbols MID1IP1
Host Species Rabbit
Immunogen Recombinant Human MID1IP1 Protein (Met1-His183).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 58526
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.