Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MID1IP1 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA571746
Description
Plays a role in the regulation of lipogenesis in liver. Up-regulates ACACA enzyme activity. Required for efficient lipid biosynthesis, including triacylglycerol, diacylglycerol and phospholipid. Involved in stabilization of microtubules (By similarity).
Specifications
MID1IP1 | |
Polyclonal | |
Unconjugated | |
MID1IP1 | |
G12-like; MIG12; S14R; STRAIT11499; THRSPL; MID1 interacting G12-like protein; MID1 interacting protein 1 (gastrulation specific G12-like); gastrulation-specific G12-like protein; mid1-interacting G12-like protein; mid1-interacting protein 1; spot 14-R; spot 14-related protein | |
Rabbit | |
selective precipitation | |
RUO | |
58526 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
20.01 mg/mL | |
PBS with 0.09% sodium azide | |
Q9NPA3 | |
MID1IP1 | |
KLH conjugated synthetic peptide between 24-51 amino acids from the N-terminal region of human MID1IP1 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction